This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01796977_g1
See other RDGA GE Assays ›
Gene Symbol:
rdgA
Gene Name:
retinal degeneration A
Gene Aliases:
CG10966, CG12660, CG15352, CG34344, CG42667, DGK, DGK2, Dmel\CG42667, Dmel_CG42667, RDGA, RdgA, dDGK2, dgk-5, x35
Chromosome Location:
Chr.X: 8907068 - 9029476 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0261549
Assay ID Dm01796977_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rdgA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042799.2 NP_001036264.2 1 - 2 677 59
NM_001103447.4 NP_001096917.1 1 - 2 677 59
NM_001201635.1 NP_001188564.1 1 - 2 677 59

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001103447.4 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

phosphatidic acid biosynthetic process
phosphatidylinositol biosynthetic process
protein kinase C-activating G-protein coupled receptor signaling pathway
phototransduction
sensory perception of sound
sensory perception of smell
rhodopsin mediated signaling pathway
deactivation of rhodopsin mediated signaling
intracellular signal transduction
thermotaxis
photoreceptor cell maintenance
diacylglycerol metabolic process
glycerolipid metabolic process
lipid phosphorylation
NAD+ kinase activity
diacylglycerol kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0