This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01793341_g1
See other LFG GE Assays ›
Gene Symbol:
Lfg TTLL3B
Gene Name:
Lifeguard
Tubulin tyrosine ligase-like 3B
Gene Aliases:
BEST:CK02213, CG3814, CK02213, Dmel\CG3814, Dmel_CG3814, LFG
CG11201, DmCG11201, Dmel\CG11201, Dmel_CG11201, TTLL3, dmTTLL3B
Chromosome Location:
Chr.2R: 12875164 - 12876966 on Build Release_6_plus_ISO1_MT
Chromosome Location:
Chr.2L: 6732866 - 6736149 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0025692
FBgn0031853
Assay ID Dm01793341_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lfg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001273998.1 NP_001260927.1 2 - 3 394 86
NM_001273999.1 NP_001260928.1 3 - 4 316 86
NM_136980.4 NP_610824.1 4 - 5 456 86
NM_165955.3 NP_725236.1 3 - 4 403 86
GenBank mRNA AY075250.1 - 3 - 4 403 86
BT015208.1 - 4 - 5 449 86
Gene Symbol: TTLL3B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BT021443.1 - 3247 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Drosophila melanogaster

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0