This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cp04182495_m1
See other INS GE Assays ›
Gene Symbol:
Ins
Gene Name:
insulin
Gene Aliases:
-
Chromosome Location:
Chr.Un|NT_176377: 12395669 - 12396848 on Build Cavpor3.0
Species:
Guinea pig
Species Specific ID (Flybase ID):
-
Assay ID Cp04182495_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ins
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172891.1 NP_001166362.1 2 - 3 237 57
XM_013150649.1 XP_013006103.1 1 - 2 1059 57
GenBank mRNA M11713.1 - 2 - 3 237 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Cavia porcellus Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0