This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05956667_g1
See other MDM4 GE Assays ›
Gene Symbol:
MDM4
Gene Name:
MDM4, p53 regulator
Gene Aliases:
-
Chromosome Location:
Chr.19: 26413339 - 26453024 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05956667_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MDM4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002760698.4 XP_002760744.1 2 - 3 387 83
XM_008985515.2 XP_008983763.1 2 - 3 391 83
XM_008985516.2 XP_008983764.1 2 - 3 249 83
XM_008985517.2 XP_008983765.1 2 - 3 401 83
XM_008985518.2 XP_008983766.1 2 - 3 254 83
XM_008985519.2 XP_008983767.1 1 - 2 119 83
XM_017966589.1 XP_017822078.1 2 - 3 237 83
XM_017966591.1 XP_017822080.1 2 - 3 670 83
XM_017966592.1 XP_017822081.1 2 - 3 386 83
XM_017966593.1 XP_017822082.1 2 - 3 251 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0