This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05954972_m1
See other AURKB GE Assays ›
Gene Symbol:
AURKB
Gene Name:
aurora kinase B
Gene Aliases:
-
Chromosome Location:
Chr.13: 50566531 - 50572531 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05954972_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AURKB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002757066.4 XP_002757112.2 7 - 8 811 95
XM_008979514.2 XP_008977762.1 7 - 8 877 95
XM_008979515.2 XP_008977763.1 7 - 8 764 95
XM_017963880.1 XP_017819369.1 2 - 3 234 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0