This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05936161_m1
See other PRLR GE Assays ›
Gene Symbol:
PRLR
Gene Name:
prolactin receptor
Gene Aliases:
-
Chromosome Location:
Chr.2: 168351831 - 168526301 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05936161_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRLR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267737.1 NP_001254666.1 4 - 5 579 70
XM_008992156.2 XP_008990404.1 6 - 7 1512 70
XM_008992158.2 XP_008990406.1 6 - 7 997 70
XM_017964581.1 XP_017820070.1 7 - 8 4345 70
XM_017964586.1 XP_017820075.1 7 - 8 4330 70
XM_017964592.1 XP_017820081.1 7 - 8 4210 70
GenBank mRNA AJ272217.1 - 4 - 5 579 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header