This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05918560_m1
See other DELE1 GE Assays ›
Gene Symbol:
DELE1
Gene Name:
DAP3 binding cell death enhancer 1
Gene Aliases:
KIAA0141
Chromosome Location:
Chr.2: 38911347 - 38927797 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05918560_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DELE1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008986670.1 XP_008984918.1 4 - 5 574 85
XM_008986676.1 XP_008984924.1 4 - 5 574 85
XM_008986688.1 XP_008984936.1 4 - 5 574 85
XM_008986693.1 XP_008984941.1 4 - 5 574 85
XM_017968447.1 XP_017823936.1 4 - 5 574 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0