This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04798926_m1
See other SH3GL3 GE Assays ›
Gene Symbol:
SH3GL3
Gene Name:
SH3 domain containing GRB2 like 3, endophilin A3
Gene Aliases:
-
Chromosome Location:
Chr.21: 23947018 - 24053895 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04798926_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SH3GL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018066197.1 XP_017921686.1 4 - 5 714 101
XM_018066198.1 XP_017921687.1 4 - 5 714 101
XM_018066199.1 XP_017921688.1 4 - 5 714 101
XM_018066200.1 XP_017921689.1 4 - 5 714 101
XM_018066201.1 XP_017921690.1 3 - 4 235 101
XM_018066202.1 XP_017921691.1 4 - 5 634 101
GenBank mRNA KA320375.1 - 4 - 5 331 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0