This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04790892_g1
See other TOGARAM1 GE Assays ›
Gene Symbol:
TOGARAM1
Gene Name:
TOG array regulator of axonemal microtubules 1
Gene Aliases:
FAM179B
Chromosome Location:
Chr.21: 54230793 - 54306820 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04790892_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TOGARAM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013973177.2 XP_013828631.2 10 - 11 3848 109
XM_013973178.2 XP_013828632.2 9 - 10 3695 109
XM_018066410.1 XP_017921899.1 10 - 11 3848 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

non-motor microtubule binding protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0