This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04776210_m1
See other IFI44 GE Assays ›
Gene Symbol:
IFI44
Gene Name:
interferon induced protein 44
Gene Aliases:
-
Chromosome Location:
Chr.3: 55187645 - 55206941 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04776210_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IFI44
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005678196.3 XP_005678253.2 4 - 5 869 73
XM_005678197.3 XP_005678254.2 4 - 5 869 73
XM_018045664.1 XP_017901153.1 4 - 5 869 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0