Assay Details
Gene Symbol: LOC106504024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044656.1 | XP_017900145.1 | 2657 | 116 |
Gene Symbol: HTT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049859.1 | XP_017905348.1 | 8139 | 116 | ||
XM_018049860.1 | XP_017905349.1 | 7799 | 116 | |||
XM_018049861.1 | XP_017905350.1 | 8091 | 116 | |||
XM_018049862.1 | XP_017905351.1 | 8007 | 116 |

More Information
Set Membership: |
![]() ![]() |
