This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04769162_mH
See other LOC102169408 GE Assays ›
Gene Symbol:
LOC102169408
Gene Name:
SEC14-like protein 4
Gene Aliases:
SEC14L4
Chromosome Location:
Chr.17: 1800664 - 1813109 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04769162_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102169408
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018060980.1 XP_017916469.1 8 - 9 735 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0