This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04760152_m1
See other LOC102187632 GE Assays ›
Gene Symbol:
LOC102187632
Gene Name:
aldehyde oxidase 1
Gene Aliases:
AOX1
Chromosome Location:
Chr.2: 46767520 - 46837515 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04760152_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102187632
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005676339.3 XP_005676396.1 7 - 8 926 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0