This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04748245_m1
See other LOC102176218 GE Assays ›
Gene Symbol:
LOC102176218
Gene Name:
zinc finger protein 2 homolog
Gene Aliases:
ZFP2, ZNF454
Chromosome Location:
Chr.7: 106324484 - 106372828 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04748245_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102176218
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018051573.1 XP_017907062.1 2 - 3 194 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_018051573.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0