Assay Details
Gene Symbol: ZNF618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052641.1 | XP_017908130.1 | 14 - 15 | 1356 | 57 | |
XM_018052642.1 | XP_017908131.1 | 14 - 15 | 1353 | 57 | ||
XM_018052643.1 | XP_017908132.1 | 13 - 14 | 1320 | 57 | ||
XM_018052644.1 | XP_017908133.1 | 13 - 14 | 1509 | 57 | ||
XM_018052645.1 | XP_017908134.1 | 13 - 14 | 1311 | 57 | ||
XM_018052646.1 | XP_017908135.1 | 13 - 14 | 1308 | 57 | ||
XM_018052648.1 | XP_017908137.1 | 13 - 14 | 1296 | 57 | ||
XM_018052649.1 | XP_017908138.1 | 12 - 13 | 1260 | 57 | ||
XM_018052650.1 | XP_017908139.1 | 12 - 13 | 1251 | 57 | ||
XM_018052651.1 | XP_017908140.1 | 12 - 13 | 1248 | 57 | ||
XM_018052652.1 | XP_017908141.1 | 13 - 14 | 1113 | 57 | ||
XM_018052653.1 | XP_017908142.1 | 12 - 13 | 1068 | 57 | ||
XM_018052654.1 | XP_017908143.1 | 11 - 12 | 987 | 57 | ||
XM_018052655.1 | XP_017908144.1 | 10 - 11 | 927 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
