This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04733492_m1
See other LOC108633807 GE Assays ›
Gene Symbol:
LOC108633807 LOC102191395
Gene Name:
D-2-hydroxyglutarate dehydrogenase, mitochondrial
D-2-hydroxyglutarate dehydrogenase, mitochondrial
Gene Aliases:
-
D2HGDH
Chromosome Location:
Chr.3: 337592 - 346705 on Build ARS1
Chromosome Location:
Chr.Un|NW_017189568: 78 - 19369 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
-
Assay ID Ch04733492_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC108633807
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018039702.1 XP_017895191.1 1702 98
XM_018039705.1 XP_017895194.1 1439 98
XM_018039709.1 XP_017895198.1 1763 98
XM_018039713.1 XP_017895202.1 1423 98
XM_018039720.1 XP_017895209.1 1535 98
Gene Symbol: LOC102191395
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018044473.1 XP_017899962.1 8 - 9 1133 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0