This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04730714_m1
See other NUDT6 GE Assays ›
Gene Symbol:
NUDT6
Gene Name:
nudix hydrolase 6
Gene Aliases:
-
Chromosome Location:
Chr.17: 36278250 - 36333689 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04730714_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013970536.2 XP_013825990.2 3 - 4 548 98
XM_018061507.1 XP_017916996.1 3 - 4 908 98
XM_018061508.1 XP_017916997.1 3 - 4 824 98
XM_018061509.1 XP_017916998.1 3 - 4 318 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

nucleotide phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0