Assay Details
Gene Symbol: KRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047712.1 | XP_017903201.1 | 4 - 5 | 1083 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Gene Symbol: KRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047712.1 | XP_017903201.1 | 4 - 5 | 1083 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |