Assay Details
Gene Symbol: FKBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679207.3 | XP_005679264.3 | 4 - 5 | 908 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: FKBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679207.3 | XP_005679264.3 | 4 - 5 | 908 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() |