This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04691006_g1
See other HSD17B8 GE Assays ›
Gene Symbol:
HSD17B8
Gene Name:
hydroxysteroid 17-beta dehydrogenase 8
Gene Aliases:
-
Chromosome Location:
Chr.23: 41289595 - 41291685 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04691006_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD17B8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018038590.1 XP_017894079.1 6 - 7 728 65
XM_018038591.1 XP_017894080.1 6 - 7 706 65
GenBank mRNA JO325702.2 - 5 - 6 529 65
JO374183.2 - 5 - 6 560 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0