This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04688540_m1
See other IMMT GE Assays ›
Gene Symbol:
IMMT
Gene Name:
inner membrane mitochondrial protein
Gene Aliases:
-
Chromosome Location:
Chr.11: 48277976 - 48316952 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04688540_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IMMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005686702.3 XP_005686759.2 1 - 2 239 57
XM_005686705.3 XP_005686762.2 1 - 2 239 57
XM_018055364.1 XP_017910853.1 1 - 2 239 57
XM_018055365.1 XP_017910854.1 1 - 2 239 57
XM_018055367.1 XP_017910856.1 1 - 2 239 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0