This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04686352_g1
See other LOC102187810 GE Assays ›
Gene Symbol:
LOC102187810 PFKL
Gene Name:
ATP-dependent 6-phosphofructokinase, liver type
phosphofructokinase, liver type
Gene Aliases:
-
-
Chromosome Location:
Chr.Un|NW_017204900: 6185 - 22324 on Build ARS1
Chromosome Location:
Chr.1: 143914619 - 143922223 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
-
Assay ID Ch04686352_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102187810
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018045395.1 XP_017900884.1 1 371 80
GenBank mRNA JO427006.1 - 19 2607 80
JO560724.2 - 378 80
JO644629.2 - 19 2551 80
Gene Symbol: PFKL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018051894.1 XP_017907383.1 358 80
GenBank mRNA JO513728.2 - 498 80
KA337450.1 - 3225 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

carbohydrate kinase kinase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0