This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04684681_m1
See other LCP1 GE Assays ›
Gene Symbol:
LCP1
Gene Name:
lymphocyte cytosolic protein 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 70260840 - 70354603 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04684681_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LCP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005687438.3 XP_005687495.2 14 - 15 1865 63
XM_005687439.3 XP_005687496.2 14 - 15 1799 63
XM_013968230.2 XP_013823684.2 14 - 15 1671 63
GenBank mRNA JO339090.2 - 3 - 4 1380 63
JO579574.1 - 2 - 3 1365 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

non-motor actin binding protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header