This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04683167_g1
See other RIPOR1 GE Assays ›
Gene Symbol:
RIPOR1
Gene Name:
RHO family interacting cell polarization regulator 1
Gene Aliases:
FAM65A
Chromosome Location:
Chr.18: 36350510 - 36399069 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04683167_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIPOR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018062151.1 XP_017917640.1 5 - 6 404 58
XM_018062152.1 XP_017917641.1 5 - 6 1534 58
XM_018062154.1 XP_017917643.1 5 - 6 404 58
XM_018062155.1 XP_017917644.1 7 - 8 1215 58
XM_018062156.1 XP_017917645.1 6 - 7 1130 58
XM_018062157.1 XP_017917646.1 5 - 6 519 58
XM_018062158.1 XP_017917647.1 5 - 6 404 58
GenBank mRNA JO430111.2 - 5 - 6 544 58
JO523991.2 - 5 - 6 543 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0