This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04682549_m1
See other BCKDHB GE Assays ›
Gene Symbol:
BCKDHB
Gene Name:
branched chain keto acid dehydrogenase E1 subunit beta
Gene Aliases:
-
Chromosome Location:
Chr.9: 7115439 - 7377325 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04682549_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCKDHB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018052910.1 XP_017908399.1 7 - 8 883 69
GenBank mRNA JO630315.2 - 4 - 5 403 69
JR114742.1 - 7 - 8 771 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0