This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04682251_m1
See other LOC102172789 GE Assays ›
Gene Symbol:
LOC102172789
Gene Name:
putative RNA-binding protein Luc7-like 2
Gene Aliases:
LUC7L2
Chromosome Location:
Chr.4: 17463910 - 17519651 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04682251_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102172789
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013963527.2 XP_013818981.2 8 - 9 877 190
XM_018046879.1 XP_017902368.1 7 - 8 1193 190
XM_018046880.1 XP_017902369.1 7 - 8 1799 190
XM_018046881.1 XP_017902370.1 8 - 9 1302 190
XM_018046882.1 XP_017902371.1 8 - 9 891 190
XM_018046883.1 XP_017902372.1 7 - 8 740 190
GenBank mRNA JO643760.2 - 8 - 9 1006 190

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0