Assay Details
Gene Symbol: MBIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066388.1 | XP_017921877.1 | 5 - 6 | 716 | 96 | |
XM_018066389.1 | XP_017921878.1 | 5 - 6 | 717 | 96 | ||
XM_018066390.1 | XP_017921879.1 | 4 - 5 | 597 | 96 | ||
XM_018066391.1 | XP_017921880.1 | 5 - 6 | 718 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
