This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04676723_mH
See other FGF9 GE Assays ›
Gene Symbol:
FGF9
Gene Name:
fibroblast growth factor 9
Gene Aliases:
-
Chromosome Location:
Chr.12: 51337881 - 51367467 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04676723_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FGF9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005687564.3 XP_005687621.1 2 - 3 1074 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0