This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04671331_m1
See other LOC102180906 GE Assays ›
Gene Symbol:
LOC102180906
Gene Name:
serpin B9
Gene Aliases:
SERPINB9
Chromosome Location:
Chr.23: 1576820 - 1612707 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04671331_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102180906
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018038978.1 XP_017894467.1 3 - 4 395 81
XM_018038979.1 XP_017894468.1 3 - 4 343 81
XM_018038980.1 XP_017894469.1 3 - 4 399 81
XM_018038981.1 XP_017894470.1 3 - 4 446 81
GenBank mRNA JO399368.2 - 4 - 5 756 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

protease inhibitor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0