This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04655391_m1
See other IGF1 GE Assays ›
Gene Symbol:
IGF1
Gene Name:
insulin like growth factor 1
Gene Aliases:
IGF-1, IGF-I, IGFI, gIGFI
Chromosome Location:
Chr.5: 64862982 - 64943171 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04655391_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285697.1 NP_001272626.1 2 - 3 226 102
XM_005680531.3 XP_005680588.1 2 - 3 504 102
XM_005680532.3 XP_005680589.1 2 - 3 506 102
XM_005680533.3 XP_005680590.1 2 - 3 512 102
XM_005680534.3 XP_005680591.1 2 - 3 509 102
XM_005680535.3 XP_005680592.1 2 - 3 317 102
XM_005680537.3 XP_005680594.1 2 - 3 487 102
XM_005680538.3 XP_005680595.1 2 - 3 496 102
XM_005680539.3 XP_005680596.1 2 - 3 493 102
GenBank mRNA D11378.1 - 2 - 3 303 102
FJ716104.1 - 2 - 3 231 102
JF896275.1 - 2 - 3 226 102
JF896276.1 - 2 - 3 226 102
JT757565.1 - 5 - 6 1807 102
JX570672.1 - 2 - 3 226 102
KJ549851.1 - 2 - 3 303 102
KJ549852.1 - 2 - 3 303 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0