This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04654189_g1
See other LOC100860959 GE Assays ›
Gene Symbol:
LOC100860959
Gene Name:
CD36-like protein
Gene Aliases:
-
Chromosome Location:
Chr.4: 79948194 - 79988090 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04654189_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100860959
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285580.1 NP_001272509.1 5 - 6 731 92
XM_013963206.2 XP_013818660.1 7 - 8 929 92
XM_013963208.2 XP_013818662.1 7 - 8 909 92
XM_013963209.2 XP_013818663.1 8 - 9 986 92
XM_013963210.2 XP_013818664.1 8 - 9 945 92
XM_013963211.2 XP_013818665.1 8 - 9 966 92
XM_013963212.2 XP_013818666.1 8 - 9 925 92
GenBank mRNA JF690774.2 - 5 - 6 731 92
JO422180.2 - 8 - 9 1331 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0