This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04294409_m1
See other CLDND1 GE Assays ›
Gene Symbol:
CLDND1
Gene Name:
claudin domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 42586816 - 42594122 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04294409_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLDND1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018057265.1 XP_017912754.1 5 - 6 676 69
XM_018057322.1 XP_017912811.1 4 - 5 659 69
XM_018057375.1 XP_017912864.1 5 - 6 701 69
XM_018057441.1 XP_017912930.1 5 - 6 711 69
GenBank mRNA JO324288.2 - 4 - 5 624 69
JO324289.2 - 4 - 5 624 69
JO344197.2 - 4 - 5 633 69
JO344198.2 - 4 - 5 633 69
JO371731.2 - 4 - 5 651 69
JO415840.2 - 1 - 2 705 69
JO415841.2 - 1 - 2 705 69
JO434961.2 - 1 - 2 821 69
JO435237.2 - 1 - 2 821 69
JO508546.2 - 1 - 2 282 69
JO533328.2 - 4 - 5 669 69
JO563306.2 - 1 - 2 373 69
JO576894.1 - 4 - 5 749 69
JT738586.1 - 1 - 2 572 69
JT763272.1 - 1 - 2 910 69
JT763273.1 - 1 - 2 910 69
KA323671.1 - 3 - 4 566 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0