This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03239715_m1
See other PQBP1 GE Assays ›
Gene Symbol:
PQBP1
Gene Name:
polyglutamine binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_017190136: 82267 - 87696 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03239715_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PQBP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005700770.3 XP_005700827.1 3 - 4 346 65
XM_005700771.3 XP_005700828.1 3 - 4 349 65
XM_005700772.3 XP_005700829.1 3 - 4 569 65
XM_013976446.2 XP_013831900.1 3 - 4 357 65
XM_013976447.2 XP_013831901.1 3 - 4 354 65
XM_013976448.2 XP_013831902.1 3 - 4 570 65
GenBank mRNA JO441986.2 - 4 - 5 670 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0