Assay Details
Gene Symbol: MTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677399.3 | XP_005677456.2 | 5 - 6 | 1257 | 82 | |
GenBank mRNA | JO392228.2 | - | 3 - 4 | 374 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
