This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03222324_m1
See other SMPX GE Assays ›
Gene Symbol:
SMPX
Gene Name:
small muscle protein X-linked
Gene Aliases:
-
Chromosome Location:
Chr.X|NW_017189516: 48310073 - 48360054 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03222324_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMPX
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005701002.3 XP_005701059.1 2 - 3 278 67
XM_013976651.2 XP_013832105.1 2 - 3 278 67
XM_018044023.1 XP_017899512.1 2 - 3 278 67
GenBank mRNA JO538189.2 - 3 - 4 645 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0