This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03213478_m1
See other AK3 GE Assays ›
Gene Symbol:
AK3
Gene Name:
adenylate kinase 3
Gene Aliases:
-
Chromosome Location:
Chr.8: 39635493 - 39656271 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03213478_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AK3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005683761.3 XP_005683818.3 3 - 4 882 66
GenBank mRNA JO425302.1 - 3 - 4 672 66
JO518218.2 - 3 - 4 371 66
JO551311.2 - 2 - 3 407 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

nucleotide kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0