This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03210996_m1
See other MTFR1L GE Assays ›
Gene Symbol:
MTFR1L
Gene Name:
mitochondrial fission regulator 1 like
Gene Aliases:
-
Chromosome Location:
Chr.2: 8696812 - 8712827 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03210996_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MTFR1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005676830.3 XP_005676887.1 3 - 4 342 71
XM_005676831.3 XP_005676888.1 3 - 4 330 71
XM_018056146.1 XP_017911635.1 3 - 4 2879 71
GenBank mRNA JO547236.2 - 4 - 5 1388 71
JT672281.1 - 7 - 8 1847 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0