This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04590005_m1
See other P4HB GE Assays ›
Gene Symbol:
P4hb
Gene Name:
prolyl 4-hydroxylase, beta polypeptide
Gene Aliases:
I79_002399, PDI, PDIA1, p58
Chromosome Location:
Chr.Un|NW_003613637: 2806917 - 2820747 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04590005_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P4hb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001246693.1 NP_001233622.1 5 - 6 893 73
XM_007654094.1 XP_007652284.1 5 - 6 779 73
GenBank mRNA AF364317.1 - 5 - 6 893 73
JI868756.1 - 5 - 6 779 73
JP041321.1 - 5 - 6 779 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0