This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04577568_m1
See other XRCC4 GE Assays ›
Gene Symbol:
Xrcc4
Gene Name:
X-ray repair complementing defective repair in Chinese hamster cells 4
Gene Aliases:
I79_011207
Chromosome Location:
Chr.Un|NW_003614041: 1186704 - 1327327 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04577568_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Xrcc4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007644720.1 XP_007642910.1 556 64
GenBank mRNA JI896032.1 - 556 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0