This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04576429_g1
See other MRPL24 GE Assays ›
Gene Symbol:
Mrpl24
Gene Name:
mitochondrial ribosomal protein L24
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003613923: 783456 - 788320 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04576429_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrpl24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003502901.2 XP_003502949.1 5 - 6 638 70
XM_007643516.1 XP_007641706.1 5 - 6 666 70
XM_007643517.1 XP_007641707.1 5 - 6 666 70
XM_007643518.1 XP_007641708.1 5 - 6 815 70
GenBank mRNA JI896166.1 - 4 - 5 567 70
JP063249.1 - 4 - 5 567 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0