Assay Details
Gene Symbol: Mtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642243.1 | XP_007640433.1 | 6 - 7 | 702 | 80 | |
GenBank mRNA | JP039223.1 | - | 6 - 7 | 609 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
