Assay Details
Gene Symbol: Gba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501588.2 | XP_003501636.1 | 3 - 4 | 445 | 57 | |
GenBank mRNA | JI880115.1 | - | 3 - 4 | 459 | 57 | |
JP053773.1 | - | 3 - 4 | 459 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
