This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04550141_m1
See other GPR56 GE Assays ›
Gene Symbol:
Gpr56
Gene Name:
G protein-coupled receptor 56
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003614770: 330472 - 360031 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04550141_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr56
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003509803.2 XP_003509851.1 8 - 9 1114 80
XM_007650448.1 XP_007648638.1 8 - 9 1342 80
XM_007650449.1 XP_007648639.1 8 - 9 1397 80
XM_007650450.1 XP_007648640.1 8 - 9 1186 80
XM_007650451.1 XP_007648641.1 8 - 9 1145 80
XM_007650452.1 XP_007648642.1 8 - 9 1125 80
XM_007650453.1 XP_007648643.1 8 - 9 1327 80
XM_007650454.1 XP_007648644.1 8 - 9 1381 80
GenBank mRNA JI870114.1 - 6 - 7 1003 80
JP062986.1 - 7 - 8 1090 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0