This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04550138_m1
See other GPR56 GE Assays ›
Gene Symbol:
Gpr56
Gene Name:
G protein-coupled receptor 56
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003614770: 330472 - 360031 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04550138_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr56
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003509803.2 XP_003509851.1 5 - 6 817 68
XM_007650448.1 XP_007648638.1 5 - 6 1045 68
XM_007650449.1 XP_007648639.1 5 - 6 1100 68
XM_007650450.1 XP_007648640.1 5 - 6 889 68
XM_007650451.1 XP_007648641.1 5 - 6 848 68
XM_007650452.1 XP_007648642.1 5 - 6 828 68
XM_007650453.1 XP_007648643.1 5 - 6 1030 68
XM_007650454.1 XP_007648644.1 5 - 6 1084 68
GenBank mRNA JI870114.1 - 3 - 4 706 68
JP062986.1 - 4 - 5 793 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header