Assay Details
Gene Symbol: Gapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496202.1 | XP_003496250.1 | 1 | 424 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Gene Symbol: Gapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496202.1 | XP_003496250.1 | 1 | 424 | 88 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |