This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04541552_m1
See other FIGF GE Assays ›
Gene Symbol:
Figf
Gene Name:
c-fos induced growth factor (vascular endothelial growth factor D)
Gene Aliases:
I79_009992
Chromosome Location:
Chr.Un|NW_003613952: 334903 - 370580 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04541552_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Figf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007643841.1 XP_007642031.1 2 - 3 1068 82
XM_007643842.1 XP_007642032.1 2 - 3 1068 82
XM_007643843.1 XP_007642033.1 2 - 3 231 82
XM_007643844.1 XP_007642034.1 2 - 3 1053 82
GenBank mRNA JI873754.1 - 2 - 3 398 82
JP043745.1 - 2 - 3 398 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0