This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04523303_m1
See other LOC100756511 GE Assays ›
Gene Symbol:
LOC100756511
Gene Name:
dual 3',5'-cyclic-AMP and -GMP phosphodiesterase 11A
Gene Aliases:
I79_007367, Pde11a
Chromosome Location:
Chr.Un|NW_003613688: 1093 - 102029 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04523303_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100756511
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003501029.2 XP_003501077.2 9 - 10 1731 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Probe spans exons

Panther Classification:

Molecular Function -

phosphodiesterase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0