This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04518974_m1
See other LOC100772034 GE Assays ›
Gene Symbol:
LOC100772034
Gene Name:
tubulin polyglutamylase TTLL13
Gene Aliases:
I79_023389, Ttll13
Chromosome Location:
Chr.Un|NW_003616436: 61121 - 77495 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04518974_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100772034
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007654401.1 XP_007652591.1 10 - 11 1612 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

microtubule or microtubule-binding cytoskeletal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0