This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04496317_m1
See other GPR126 GE Assays ›
Gene Symbol:
Gpr126
Gene Name:
G protein-coupled receptor 126
Gene Aliases:
I79_007561
Chromosome Location:
Chr.Un|NW_003613836: 81529 - 210402 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04496317_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr126
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007641869.1 XP_007640059.1 9 - 10 1650 77
XM_007641870.1 XP_007640060.1 8 - 9 1566 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0