Assay Details
Gene Symbol: Thpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509619.2 | XP_003509667.1 | 2 - 3 | 228 | 71 | |
XM_007650217.1 | XP_007648407.1 | 2 - 3 | 228 | 71 | ||
XM_007650218.1 | XP_007648408.1 | 2 - 3 | 228 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
